Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD62L Mouse, anti-Rat, R718, Clone: HRL1, BD OptiBuild™

Catalog No. BDB752003 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752003 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752003 Supplier BD Biosciences Supplier No. 752003
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HRL1 monoclonal antibody specifically binds to L-selectin (LECAM-1, CD62L), which is detected on a small percentage of thymocytes and on most neutrophils and peripheral lymphocytes. CD62L is a 62-kDa (on neutrophils) or 65-kDa (on lymphocytes) receptor, with lectin-like and epidermal growth factor-like domains, which binds to sialylated oligosaccharide determinants on high endothelial venules (HEV) in peripheral lymph nodes. This member of the selectin adhesion molecule family appears to be required for lymphocyte homing to peripheral lymph nodes and to contribute to neutrophil emigration at inflammatory sites. L-selectin is rapidly shed from lymphocytes and neutrophils upon cell activation. In the mouse, the level of CD62L expression distinguishes naive CD4+T cells from effector/memory T helper cells. HRL1 antibody inhibits the ligand-binding activity of Lselectin in in vitro assays and slows in vivo leukocyte rolling on microvascular endothelium.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD62L
Applications Flow Cytometry
Classification Monoclonal
Clone HRL1
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.