Learn More
CD62P Mouse anti-Human, APC, Clone: AK-4, BD
Mouse Monoclonal Antibody
$212.00 - $430.50
Specifications
| Antigen | CD62P |
|---|---|
| Clone | AK-4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
BDB561920
|
BD Biosciences
561920 |
25 Tests |
Each for $212.00
|
|
|||||
|
BDB550888
|
BD Biosciences
550888 |
100 Tests |
Each for $430.50
|
|
|||||
Description
The AK-4 monoclonal antibody specifically binds to CD62P. CD62P is a 140 kDa type I transmembrane glycoprotein that is also known as P-Selectin, Platelet activation-dependent granule membrane protein (PADGEM), or GMP-140. P-Selectin is stored in the α-granules of platelets and the Weibel-Palade bodies of endothelial cells, and is rapidly transported to the plasma membrane upon activation. P-Selectin is thought to mediate the initial adhesive interactions of neutrophils and monocytes with endothelium in inflammatory responses, and of activated platelets to neutrophils and monocytes in hemostasis.
Specifications
| CD62P | |
| Flow Cytometry | |
| APC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| AK-4 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity Purified | |
| P-Selectin; GMP-140; PADGEM |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.