Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD62P Mouse, anti-Human, R718, Clone: AK-4, BD OptiBuild™

Catalog No. BDB751983 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751983 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751983 Supplier BD Biosciences Supplier No. 751983
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The AK-4 monoclonal antibody specifically binds to CD62P. CD62P is a 140 kDa type I transmembrane glycoprotein that is also known as P-Selectin, Platelet activation-dependent granule membrane protein (PADGEM), or Granule membrane protein-140 (GMP-140). CD62P is expressed by megakaryocytes, activated platelets and activated endothelium. P-Selectin is stored in the α-granules of platelets and the Weibel-Palade bodies of endothelial cells, and is rapidly transported to the plasma membrane upon activation. P-Selectin is thought to mediate the initial adhesive interactions of neutrophils and monocytes with endothelium in inflammatory responses, and of activated platelets to neutrophils and monocytes in hemostasis.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD62P
Applications Flow Cytometry
Classification Monoclonal
Clone AK-4
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.