Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD63 Antibody (7G4.2E8), Allophycocyanin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP236567APC
Description
CD63 Monoclonal specifically detects CD63 in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Intracellular).Specifications
CD63 | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1lysosome-associated membrane glycoprotein 3, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, TSPAN30granulophysin | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry | |
7G4.2E8 | |
Flow Cytometry, Flow (Intracellular) | |
P41731 | |
CD63 | |
Partial recombinant mouse CD63 protein (between amino acids 100-250) [UniProt P41731] | |
0.1 mL | |
Autophagy, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis | |
967 | |
Store at 4C in the dark. | |
IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction