Learn More
CD63 Antibody (7G4.2E8) - BSA Free, Novus Biologicals™
SDP
Rat Monoclonal Antibody has been used in 2 publications
Supplier:  Novus Biologicals NBP236567
 View more versions of this product
Catalog No. NBP236567
Only null left
Add to Cart
Description
Description
CD63 Monoclonal specifically detects CD63 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Flow (Intracellular).
Specifications
Specifications
CD63
Monoclonal
1.0 mg/mL
Western Blot 2.0 ug/ml, Flow Cytometry 2 ug 1 million cells, Flow (Intracellular) 2 ug 1 million cells
P41731
CD63
Partial recombinant mouse CD63 protein (between amino acids 100-250) [UniProt P41731]
0.1 mg
Autophagy, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis
967
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2a λ
Western Blot, Flow Cytometry
7G4.2E8
Unconjugated
PBS with 0.05% Sodium Azide
CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1lysosome-associated membrane glycoprotein 3, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, TSPAN30granulophysin
Rat
Protein G purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only