Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD63 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$332.50 - $545.00
Specifications
Antigen | CD63 |
---|---|
Dilution | Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL |
Applications | Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CD63 Polyclonal antibody specifically detects CD63 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
CD63 | |
Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Autophagy, Cancer, Cardiovascular Biology, Cellular Signaling, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis | |
PBS, pH 7.2, 40% glycerol | |
967 | |
IgG | |
Affinity purified |
Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL | |
Polyclonal | |
Purified | |
RUO | |
Human | |
CD_antigen: CD63, CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, lysosome-associated membrane glycoprotein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: VGAQLVLSQTIIQGATPGSL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title