Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD63 Antibody (OTI5E5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246167
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
CD63 Monoclonal specifically detects CD63 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CD63 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1lysosome-associated membrane glycoprotein 3, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, TSPAN30granulophysin | |
Mouse | |
25.5 kDa | |
0.1 mL | |
Autophagy, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis | |
967 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI5E5 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
CD63 | |
Full length human recombinant protein of human CD63 (NP_001771) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction