Learn More
CD68 Mouse anti-Human, Clone: CD68/G2, Abnova™
Mouse monoclonal antibody raised against full length recombinant human CD68.
Supplier: Abnova Corporation MAB13483
Description
This gene encodes a 110-kD transmembrane glycoprotein that is highly expressed by human monocytes and tissue macrophages. It is a member of the lysosomal/endosomal-associated membrane glycoprotein (LAMP) family. The protein primarily localizes to lysosomes and endosomes with a smaller fraction circulating to the cell surface. It is a type I integral membrane protein with a heavily glycosylated extracellular domain and binds to tissue- and organ-specific lectins or selectins. The protein is also a member of the scavenger receptor family. Scavenger receptors typically function to clear cellular debris, promote phagocytosis, and mediate the recruitment and activation of macrophages. Alternative splicing results in multiple transcripts encoding different isoforms. [provided by RefSeq]
Specifications
| CD68 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| DKFZp686M18236/GP110/SCARD1 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 968 | |
| Store at 4°C. | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| CD68/G2 | |
| Mouse monoclonal antibody raised against full length recombinant human CD68. | |
| In 10mM PBS (0.05% BSA, 0.05% sodium azide). | |
| CD68 | |
| Recombinant protein corresponding to full length human CD68. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.