Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD63 Mouse anti-Human, Clone: TS63, Novus Biologicals™

Mouse Monoclonal Antibody
$469.50 - $997.50
Specifications
Antigen | CD63 |
---|---|
Clone | TS63 |
Concentration | 1 mg/mL |
Dilution | Western Blot, Flow Cytometry, Immunoprecipitation |
Applications | Western Blot, Flow Cytometry, Immunoprecipitation |
Description
CD63 Monoclonal antibody specifically detects CD63 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunoprecipitationSpecifications
CD63 | |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Autophagy, Cancer, Cardiovascular Biology, Cellular Signaling, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis | |
PBS. Sterile-filtered through 0.22 um. | |
967 | |
IgG1 κ | |
Affinity purified | |
Antibody |
TS63 | |
Western Blot, Flow Cytometry, Immunoprecipitation | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD_antigen: CD63, CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, lysosome-associated membrane glycoprotein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30 | |
Jurkat and HEL cell lines | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title