Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD64[a,b] Mouse anti-Mouse, BV605, Clone: X54-5/7.1 (also known as X54-5/7.1), BD Horizon™

Catalog No. BDB569299 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569299 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB569299 Supplier BD Biosciences Supplier No. 569299
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD64[a,b] Monoclonal antibody specifically detects CD64[a,b] in Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD64[a,b]
Applications Flow Cytometry
Classification Monoclonal
Clone X54-5/7.1 (also known as X54-5/7.1)
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 605
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Fcgr1; FcRI; Fcg1; Fc-gamma RI; Fc receptor IgG high affinity I; IGGHAFC
Host Species Mouse
Immunogen Mouse CD64 a Alloantigen
Purification Method Affinity Chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14129
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.