Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD64[a,b] Mouse anti-Mouse, BV786, Clone: X54-5/7.1 (also known as X54-5/7.1), BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569507
Description
CD64[a,b] Monoclonal antibody specifically detects CD64[a,b] in Mouse samples. It is validated for Flow cytometry
Specifications
CD64[a,b] | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
14129 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG1 κ |
Flow Cytometry | |
X54-5/7.1 (also known as X54-5/7.1) | |
Brilliant Violet 786 | |
Fcgr1; FcRI; Fcg1; Fc-gamma RI; Fc receptor IgG high affinity I; IGGHAFC | |
Mouse CD64 a Alloantigen | |
50 μg | |
Primary | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction