Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD64 (FCGR1) Monoclonal Antibody (10.1), Functional Grade, In Vivo PLATINUM™, Leinco Technologies™
SDP

Catalog No. 502544148
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4148 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4148 Supplier Leinco Technologies Inc Supplier No. I2015100MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specificity: Clone 10.1 recognizes the alpha subunit of human FCGR1. Endotoxin Level: ≤ 0.5 EU/mg as determined by the LAL method. Purity: ≥98% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Pathogen Testing: To protect mouse colonies from infection by pathogens and to assure that experimental preclinical data is not affected by such pathogens, all of Leinco Purified Functional PLATINUM™ antibodies are tested and guaranteed to be negative for all pathogens in the IDEXX IMPACT I Mouse Profile. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.

CD64 (FCGR1) Monoclonal antibody specifically detects CD64 (FCGR1) in Human samples. It is validated for Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Inhibition Assays

Specifications

Antigen CD64 (FCGR1)
Applications Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Inhibition Assays
Classification Monoclonal
Clone 10.1
Concentration ≥5.0 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2-7.4
Gene FCGR1A
Gene Accession No. P12314
Gene Alias AI323638; AV092959; CD64; CD64 antigen; CD64A; Fc fragment of IgG high affinity Ib receptor; Fc fragment of IgG receptor Ia; Fc fragment of IgG, high affinity Ia, receptor (CD64); Fc fragment of IgG, high affinity Ia, receptor for (CD64); Fc fragment of IgG, high affinity Ib, receptor for (CD64); Fc gamma receptor; Fc gamma receptor Ia; Fc receptor, IgG, high affinity I; Fcg1; Fc-gamma receptor 1; Fc-gamma receptor I A1; Fc-gamma RI; Fc-gamma RIA; FcgammaRI; FcgammaRIa; Fcgr1; FCGR1A; Fcgr1b; FcgRI; FCRI; FLJ18345; high affinity IgG Fc receptor, subunit beta; high affinity immunoglobulin gamma Fc receptor I; high-affinity immunoglobulin gamma Fc receptor I; IGFR1; IgG Fc receptor I; IgG high affinity Fc receptor; IGGHAFC; RP11-196G18.2
Host Species Mouse
Immunogen Rheumatoid synovial fluid cells and fibronectin purified human monocytes
Purification Method Protein A/G
Quantity 100 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2209
Target Species Human
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.