Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD64 Monoclonal Antibody (10.1)
GREENER_CHOICE

Catalog No. PIMA110270
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA110270 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA110270 Supplier Invitrogen™ Supplier No. MA110270
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes an extracellular epitope on CD64/FcgammaRI, a 72 kDa single chain type I glycoprotein, that is expressed on monocytes/macrophages, dendritic cells, and activated granulocytes. The epitope is sensitive to formalin fixation.

CD64 (FCGR1A) encodes a protein that plays an important role in the immune response. CD64 is a high-affinity Fc-gamma receptor and is one of three related gene family members located on chromosome 1. CD64 plays an important role in the immune response and its dysfunction has been studied in cervical adenitis and tetrasomy 21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD64
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 10.1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene FCGR1A
Gene Accession No. P12314
Gene Alias AI323638; AV092959; CD64; CD64 antigen; CD64A; Fc fragment of IgG high affinity Ib receptor; Fc fragment of IgG receptor Ia; Fc fragment of IgG, high affinity Ia, receptor (CD64); Fc fragment of IgG, high affinity Ia, receptor for (CD64); Fc fragment of IgG, high affinity Ib, receptor for (CD64); Fc gamma receptor; Fc gamma receptor Ia; Fc receptor, IgG, high affinity I; Fcg1; Fc-gamma receptor 1; Fc-gamma receptor I A1; Fc-gamma RI; Fc-gamma RIA; FcgammaRI; FcgammaRIa; Fcgr1; FCGR1A; Fcgr1b; FcgRI; FCRI; FLJ18345; high affinity IgG Fc receptor, subunit beta; high affinity immunoglobulin gamma Fc receptor I; high-affinity immunoglobulin gamma Fc receptor I; IGFR1; IgG Fc receptor I; IgG high affinity Fc receptor; IGGHAFC; RP11-196G18.2
Gene Symbols FCGR1A
Host Species Mouse
Immunogen Rheumatoid synovial fluid cells and fibronectin purified human monocytes.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2209
Target Species Human, Monkey
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.