Learn More
Invitrogen™ CD64 Monoclonal Antibody (10.1), Alexa Fluor™ 647

Description
This antibody recognizes an extracellular epitope on CD64/FcgammaRI, a 72 kDa single chain type I glycoprotein, that is expressed on monocytes/macrophages, dendritic cells, and activated granulocytes. The epitope is sensitive to formalin fixation.
Specifications
Specifications
| Antigen | CD64 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 10.1 |
| Concentration | 0.130 mg/mL |
| Conjugate | Alexa Fluor 647 |
| Formulation | PBS with 0.2% BSA and 15mM sodium azide |
| Gene | FCGR1A |
| Gene Accession No. | P12314 |
| Gene Alias | AI323638; AV092959; CD64; CD64 antigen; CD64A; Fc fragment of IgG high affinity Ib receptor; Fc fragment of IgG receptor Ia; Fc fragment of IgG, high affinity Ia, receptor (CD64); Fc fragment of IgG, high affinity Ia, receptor for (CD64); Fc fragment of IgG, high affinity Ib, receptor for (CD64); Fc gamma receptor; Fc gamma receptor Ia; Fc receptor, IgG, high affinity I; Fcg1; Fc-gamma receptor 1; Fc-gamma receptor I A1; Fc-gamma RI; Fc-gamma RIA; FcgammaRI; FcgammaRIa; Fcgr1; FCGR1A; Fcgr1b; FcgRI; FCRI; FLJ18345; high affinity IgG Fc receptor, subunit beta; high affinity immunoglobulin gamma Fc receptor I; high-affinity immunoglobulin gamma Fc receptor I; IGFR1; IgG Fc receptor I; IgG high affinity Fc receptor; IGGHAFC; RP11-196G18.2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.