Learn More
Invitrogen™ CD64 Monoclonal Antibody (10.1), PE

Mouse Monoclonal Antibody
$572.00
Specifications
Antigen | CD64 |
---|---|
Clone | 10.1 |
Concentration | Conc. Not Determined |
Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA516436
![]() |
Invitrogen™
MA516436 |
100 Tests |
Each of 1 for $572.00
|
|
|||||
Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD64 antibody, clone 10.1 recognizes the human CD64 cell surface antigen, a ~75 kDa glycoprotein expressed by monocytes.
CD64 (FCGR1A) encodes a protein that plays an important role in the immune response. CD64 is a high-affinity Fc-gamma receptor and is one of three related gene family members located on chromosome 1. CD64 plays an important role in the immune response and its dysfunction has been studied in cervical adenitis and tetrasomy 21.Specifications
CD64 | |
Conc. Not Determined | |
Flow Cytometry | |
PE | |
Mouse | |
RUO | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
P12314 | |
2209 | |
Human monocytes. | |
Antibody |
10.1 | |
4°C, store in dark, DO NOT FREEZE! | |
Monoclonal | |
Lyophilized | |
IgG1 | |
Human, Monkey | |
FCGR1A | |
AI323638; AV092959; CD64; CD64 antigen; CD64A; Fc fragment of IgG high affinity Ib receptor; Fc fragment of IgG receptor Ia; Fc fragment of IgG, high affinity Ia, receptor (CD64); Fc fragment of IgG, high affinity Ia, receptor for (CD64); Fc fragment of IgG, high affinity Ib, receptor for (CD64); Fc gamma receptor; Fc gamma receptor Ia; Fc receptor, IgG, high affinity I; Fcg1; Fc-gamma receptor 1; Fc-gamma receptor I A1; Fc-gamma RI; Fc-gamma RIA; FcgammaRI; FcgammaRIa; Fcgr1; FCGR1A; Fcgr1b; FcgRI; FCRI; FLJ18345; high affinity IgG Fc receptor, subunit beta; high affinity immunoglobulin gamma Fc receptor I; high-affinity immunoglobulin gamma Fc receptor I; IGFR1; IgG Fc receptor I; IgG high affinity Fc receptor; IGGHAFC; RP11-196G18.2 | |
FCGR1A | |
Primary | |
Protein G |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.