Learn More
Invitrogen™ CD64 Monoclonal Antibody (10.1), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ 25064942

Description
Description: The 10.1 monoclonal antibody reacts with human CD64 (FcRI), a 75 kDa type I transmembrane protein. CD64 is the high affinity receptor for IgG and is involved in antibody-dependent cell-mediated cytotoxicity (ADCC), phagocytosis, and regulation of cytokine production. Monocytes and macrophages express CD64, while mature granulocytes and lymphocytes are negative. Applications Reported: This 10.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 10.1 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222-49) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333-57) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.
CD64 (FCGR1A) encodes a protein that plays an important role in the immune response. CD64 is a high-affinity Fc-gamma receptor and is one of three related gene family members located on chromosome 1. CD64 plays an important role in the immune response and its dysfunction has been studied in cervical adenitis and tetrasomy 21.
Specifications
CD64 | |
Monoclonal | |
5 μL/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P12314 | |
FCGR1A | |
Affinity chromatography | |
RUO | |
2209 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
10.1 | |
PE-Cyanine7 | |
FCGR1A | |
AI323638; AV092959; CD64; CD64 antigen; CD64A; Fc fragment of IgG high affinity Ib receptor; Fc fragment of IgG receptor Ia; Fc fragment of IgG, high affinity Ia, receptor (CD64); Fc fragment of IgG, high affinity Ia, receptor for (CD64); Fc fragment of IgG, high affinity Ib, receptor for (CD64); Fc gamma receptor; Fc gamma receptor Ia; Fc receptor, IgG, high affinity I; Fcg1; Fc-gamma receptor 1; Fc-gamma receptor I A1; Fc-gamma RI; Fc-gamma RIA; FcgammaRI; FcgammaRIa; Fcgr1; FCGR1A; Fcgr1b; FcgRI; FCRI; FLJ18345; high affinity IgG Fc receptor, subunit beta; high affinity immunoglobulin gamma Fc receptor I; high-affinity immunoglobulin gamma Fc receptor I; IGFR1; IgG Fc receptor I; IgG high affinity Fc receptor; IGGHAFC; RP11-196G18.2 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.