Learn More
CD64 Mouse anti-Human, FITC, Clone: 10.1, BD
Mouse Monoclonal Antibody
$104.00 - $230.00
Specifications
Antigen | CD64 |
---|---|
Clone | 10.1 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Description
The 10.1 monoclonal antibody specifically binds to CD64, a 72 kDa type I transmembrane glycoprotein that is a high affinity receptor for human IgG (FcγRI), especially the IgG1 and IgG3 subclasses. CD64 is expressed on monocytes, macrophages, dendritic cells, granulocytes activated with interferon-gamma and early myeloid lineage cells. CD64 associates with a signaling FcRγ homodimer to form the functional high affinity FcγRI complex. CD64 functions in both innate and adaptive immune responses and mediates endocytosis, phagocytosis, antigen presentation, antibody-dependent cellular toxicity, cytokine release and superoxide generation.
Specifications
CD64 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified |
10.1 | |
Monoclonal | |
Mouse | |
Human | |
Human Rheumatoid synovial fluid cells and fibronectin-purified monocytes | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.