Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD66 Monoclonal Antibody (CD66a-B1.1), Super Bright™ 702, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$461.00
Specifications
Antigen | CD66 |
---|---|
Clone | CD66a-B1.1 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
67-066-842
![]() |
Invitrogen™
67066842 |
100 Tests |
Each of 1 for $461.00
|
|
|||||
Description
Mouse Monoclonal Antibody
The CD66 molecules are 180-200 kDa glycoproteins of carcinoembryonic antigen family. They are present on all blood granulocytes and some tissue macrophages, but are absent from other hematopoietic cells. The expression of CD66 increases significantly on granulocytes upon their activation. CD66a (BGP1, CEACAM1) and CD66d (CGM1, CEACAM3), as well as CEACAM2 and 4, are transmembrane proteins, whereas CD66c (CEAL, NCA, CEACAM6) and CD66e (CEA, CEACAM5), as well as CEACAM7 and 8, are anchored to the plasma membrane by C-terminal glycosylphosphatidylinositol (GPI) lipid moiety.Specifications
CD66 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
1048, 1084, 1089, 4680, 634 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
CD66a-B1.1 | |
Flow Cytometry | |
Super Bright 702 | |
Mouse | |
Human | |
O75871, P06731, P13688, P40198, P40199 | |
CEACAM1, CEACAM3, CEACAM4, CEACAM5, CEACAM6 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title