Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD66a (CEACAM1a) Mouse anti-Mouse, BB700, Clone: CC1 (also known as MAb-CCl; mAb CC1), BD Biosciences™

Catalog No. BDB750877 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB750877 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB750877 Supplier BD Biosciences Supplier No. 750877
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD66a (CEACAM1a) Monoclonal antibody specifically detects CD66a (CEACAM1a) in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD66a (CEACAM1a)
Applications Flow Cytometry
Classification Monoclonal
Clone CC1 (also known as MAb-CCl; mAb CC1)
Conjugate Brilliant Blue 700 (BB700)
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CEACAM1[a]; Ceacam1; BGP-1; Bgp; MHV-R; MHVR
Host Species Mouse
Immunogen BALB/c mouse intestinal brush border membrane proteins
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.