Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD66c Mouse anti-Human, Brilliant Violet 786, Clone: B6.2/CD66, BD Optibuild™

Catalog No. BDB742688 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742688 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742688 Supplier BD Biosciences Supplier No. 742688
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The B6.2 monoclonal antibody reacts with a glycosylphosphatidylinositol-anchored glycoprotein present on granulocytes. Antibody B6.2 was studied as recognizing CD66c in the VI Human Leukocyte Differentiation Antigen workshop. CD66 antigens also known as the carcinoembryonic antigen (CEA) family of molecules, are closely related to the immunoglobulin super family of glycoproteins. Studies on CD66 molecules suggest a potential adhesion function in vivo. These molecules exhibit both homophilic and heterophilic adhesion. CEA family members may be involved in transmembrane signalling and activation of neutrophils. This clone has been found to be N-terminal domain reactive, reacted preferentially with the native protein and were conformationally dependent

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD66c
Classification Monoclonal
Clone B6.2/CD66
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CEACAM6, Carcinoembryonic antigen-related cell adhesion molecule 6, NCA
Host Species Mouse
Immunogen Human Breast Carcinoma Extracts
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.