Learn More
CD66c Mouse anti-Human, PE, Clone: KOR-SA3544, eBioscience
Mouse Monoclonal Antibody
Manufacturer: Invitrogen 12066741
Description
Description: This KOR-SA3544 monoclonal antibody reacts with human CD66c (also known as CEACAM6), which is one of the splice variants of CD66. CD66c is expressed on endothelial and epithelial cells, as well as on neutrophils and monocytes. Myeloma and cancer cells, such as B-cell acute lymphoblastic lymphomas, also express CD66c. Moreover, expression of this receptor can be induced on T cells, B cells, and CD16-negative NK cells. Like the other CD66 splice variants, CD66c is involved in cell adhesion and growth, neutrophil activation, and signaling. Moreover, expression of CD66c in leukemic cells has been correlated with BCR/ABL1 rearrangement. Finally, studies have shown that the CD66 variants interact with each other homo- and heterotypically. Applications Reported: This KOR-SA3544 antibody has been reported for use in flow cytometric analysis. Applications Tested: This KOR-SA3544 antibody has been pre-titrated and tested by flow cytometry analysis of normal human peripheral blood cells. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 µm post-manufacturing filtered.
CEACAM1 (BGP/CD66a) is a transmembrane glycoprotein which belongs to the carcinoembryonic antigen (CEA) gene family. It is expressed on cells of epithelial and myeloid origin and mediates homophilic intercellular interactions that influence cellular growth, immune cell activation, and tissue morphogenesis. CEACAM1 is a putative tumour suppressor based on diminished expression in some aggressive types of cancer cells. The anti-tumour effect may be due to inhibition of tumour angiogenesis, possibly by increased secretion of anti-angiogenic molecules from the cells. Like all members of the CEACAM family, it consists of a single N domain, with structural homology to the immunoglobulin variable domains, followed by two immunoglobulin constant-like A (A1, A2) and one B domain. While the N, A1 and B domains can also be found in other CEA-family members, the A2 domain of CEACAM1 differs from those found in other CEACAM.Specifications
CD66c | |
Monoclonal | |
5 μL/Test | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P40199 | |
CD66c, CEAL, NCA, carcinoembryonic antigen-related cell adhesion molecule 6, non-specific crossreacting antigen, normal cross-reacting antigen | |
Mouse | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Flow Cytometry | |
KOR-SA3544 | |
PE | |
CEACAM6 | |
Liquid | |
CEACAM6 | |
IgG1, kappa | |
25 Tests | |
4° C, store in dark, DO NOT FREEZE! | |
Primary | |
4680 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.