Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD66c Monoclonal Antibody (KOR-SA3544), PE, eBioscience™, Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

$153.00 - $359.00

Specifications

Antigen CD66c
Clone KOR-SA3544
Concentration 5 μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-245-273
Encompass_Preferred
View Documents
Invitrogen™
12066741
25 Tests
Each for $153.00
Only null left
Edge
Add to Cart
 
50-245-274
GREENER_CHOICE
Encompass_Preferred
View Documents
Invitrogen™
12066742
100 Tests
Each for $359.00
Only null left
Add to Cart
 
Description

Description

Description: This KOR-SA3544 monoclonal antibody reacts with human CD66c (also known as CEACAM6), which is one of the splice variants of CD66. CD66c is expressed on endothelial and epithelial cells, as well as on neutrophils and monocytes. Myeloma and cancer cells, such as B-cell acute lymphoblastic lymphomas, also express CD66c. Moreover, expression of this receptor can be induced on T cells, B cells, and CD16-negative NK cells. Like the other CD66 splice variants, CD66c is involved in cell adhesion and growth, neutrophil activation, and signaling. Moreover, expression of CD66c in leukemic cells has been correlated with BCR/ABL1 rearrangement. Finally, studies have shown that the CD66 variants interact with each other homo- and heterotypically. Applications Reported: This KOR-SA3544 antibody has been reported for use in flow cytometric analysis. Applications Tested: This KOR-SA3544 antibody has been pre-titrated and tested by flow cytometry analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

CEA Cell Adhesion Molecule 6 (CEACAM6) belongs to the carcinoembryonic antigen (CEA) family whose members are glycosyl phosphatidyl inositol (GPI) anchored cell surface glycoproteins. Members of this family play a role in cell adhesion and are widely used as tumor markers in serum immunoassay determinations of carcinoma. CEACAM6 affects the sensitivity of tumor cells to adenovirus infection. The acts as a receptor for adherent-invasive E. coli adhesion to the surface of ileal epithelial cells in patients with Crohn's disease.
Specifications

Specifications

CD66c
5 μL/Test
Monoclonal
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
BGP; BGP-1; carcinoembryonic antigen related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific cross reacting antigen); CD66a; CD66c; CD66d; CD66e; CEA; CEACAM-1; CEACAM-3; CEACAM-5; CEACAM6; CEACAM-6; CEAL; CGM1; Cluster of Differentiation 66c; NCA; Non-specific crossreacting antigen; Normal cross-reacting antigen; pan CEACAM
CEACAM6
Primary
4°C, store in dark, DO NOT FREEZE!
CEACAM6
KOR-SA3544
Flow Cytometry
PE
Mouse
Human
P40199
4680
IgG1 κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.