Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD68 Mouse anti-Human, R718, Clone: Y1/82A, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 568069
This item is not returnable.
View return policy
Description
CD68 Monoclonal antibody specifically detects CD68 in Human samples. It is validated for Intracellular staining (Flow Cytometry)
Specifications
CD68 | |
Monoclonal | |
R718 | |
GP110; Macrosialin; SCARD1; Scavenger receptor class D, member 1 | |
PHA-stimulated Human PBMC | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
Y1/82A | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
IgG2b κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction