Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD68 Rabbit anti-Human, Unconjugated, Clone: C68/2908R, Abnova™

Catalog No. 89339716 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-716 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-716 Supplier Abnova Corporation Supplier No. RAB00677
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

CD68 Monoclonal antibody specifically detects CD68 in Human samples. It is validated for WB-Ti, IHC-P.

Specifications

Antigen CD68
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone C68/2908R
Conjugate Unconjugated
Description CD68 molecule
Gene CD68
Gene Alias DKFZp686M18236, GP110, SCARD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to full-length human CD68
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 968
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.