Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD68 Rat anti-Mouse, BV421, Clone: FA/11 (also known as FA-11, FA11), BD Horizon™
Rat Monoclonal Antibody
$204.00 - $442.00
Specifications
| Antigen | CD68 |
|---|---|
| Clone | FA/11 (also known as FA-11, FA11) |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
CD68 Monoclonal antibody specifically detects CD68 in Mouse samples. It is validated for Flow Cytometry.Specifications
| CD68 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG2a κ | |
| Refrigerate (2 to 8°C) |
| FA/11 (also known as FA-11, FA11) | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| Rat | |
| Mouse | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title