Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD68 Recombinant Mouse Monoclonal Antibody (KP-1)

Mouse Recombinant Monoclonal Antibody
$173.10 - $523.50
Specifications
Antigen | CD68 |
---|---|
Clone | KP-1 |
Concentration | 50 μg/mL |
Content And Storage | 4°C |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544195
![]() |
Invitrogen™
MA544195 |
100 μL |
Each for $523.50
|
|
|||||
PIMA544196
|
Invitrogen™
MA544196 |
10 μL |
Each for $173.10
|
|
|||||
Description
CD68 Recombinant Monoclonal Antibody for IHC (P), Flow
CD68 (Macrosialin) is a 110 kDa integral membrane glycoprotein predominantly expressed on the intracellular lysomsomes of monocytes and macrophages and to a lesser extent by dendritic cells and peripheral blood granulocytes. Also, CD68 could play a role in phagocytic activities of tissue macrophages, both in intracellular lysosomal metabolism and extracellular cell-cell and cell-pathogen interactions. CD68 is expressed by interdigitating reticulum cells in tonsil and some histiocytic lymphoma or histiocytosis, acute myeloid leukemia (AML), and granulocytic sarcoma. Elevated expression of CD68 has been demonstrated on CD34+ cells in various human malignancies, including several Acute Myeloid Leukemia studies.Specifications
CD68 | |
50 μg/mL | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 0.1% BSA and 0.09% sodium azide; pH 6.8-7.4 | |
P34810 | |
968 | |
Lysosomal Fraction Of Human Lung Macrophages. | |
Antibody |
KP-1 | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG1 | |
Human | |
Cd68 | |
CD68; CD68 antigen; CD68 molecule; GP110; Lamp4; macrophage antigen CD68; macrosialin; Scard1; scavenger receptor class D, member 1; similar to CD68 antigen | |
Cd68 | |
Primary | |
Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title