Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD68/SR-D1 Antibody (2449C) [DyLight 488], Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB10114K
Description
CD68/SR-D1 Monoclonal antibody specifically detects CD68/SR-D1 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOFSpecifications
CD68/SR-D1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
2449C | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
CD_antigen: CD68, CD68, CD68 antigen, CD68 molecule, DKFZp686M18236, GP110, LAMP4, macrophage antigen CD68, macrosialin, SCARD1, scavenger receptor class D, member 1 | |
Chinese Hamster Ovary cell line, CHO-derived mouse CD68, Asp21-Pro282, Accession # P31996 | |
0.1 mL | |
Cell Biology, Cytokine Research, Immunology, Microglia Markers | |
968 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction