Learn More
CD68/SR-D1 Antibody (2449D), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB101141MFV450
Description
CD68/SR-D1 Monoclonal antibody specifically detects CD68/SR-D1 in Mouse samples. It is validated for Immunohistochemistry, Flow Cytometry, CyTOFSpecifications
CD68/SR-D1 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
0.1 mL | |
Cell Biology, Cytokine Research, Immunology, Microglia Markers | |
968 | |
Store at 4C in the dark. |
Immunohistochemistry, Flow Cytometry, CyTOF | |
2449D | |
Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
CD_antigen: CD68, CD68, CD68 antigen, CD68 molecule, DKFZp686M18236, GP110, LAMP4, macrophage antigen CD68, macrosialin, SCARD1, scavenger receptor class D, member 1 | |
Chinese Hamster Ovary cell line, CHO-derived mouse CD68, Asp21-Pro282, Accession # P31996 | |
RUO | |
Primary | |
Mouse | |