Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD68/SR-D1 Antibody (ED1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 12 publications
$215.00 - $606.50
Specifications
Antigen | CD68/SR-D1 |
---|---|
Clone | ED1 |
Concentration | 1.0 mg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, Immunoprecipitation, Immunocytochemistry |
Classification | Monoclonal |
Description
CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Human, Mouse, Rat, Bovine, Feline, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Radioimmunoassay, Flow (Intracellular).Specifications
CD68/SR-D1 | |
1.0 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Q4FZY1 | |
968 | |
IgG1 | |
Protein A or G purified |
ED1 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, Immunoprecipitation, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Primate, Feline, Bovine, Mouse, Human, Rat, Monkey | |
CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1 | |
CD68 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title