Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD68/SR-D1 Antibody (KP1), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100683B
Description
CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy.Specifications
CD68/SR-D1 | |
Monoclonal | |
Biotin | |
PBS with 0.05% sodium azide | |
CD68 | |
This CD68/SR-D1 Antibody (KP1) was developed against subcellular fraction of human alveolar macrophages. | |
0.1 mL | |
Primary | |
This CD68/SR-D1 Antibody (KP1) is specific to macrophages in a wide variety of human tissues. It reacts with myeloid precursors and peripheral blood granulocytes. It also stains a cell population known as Plasmacytoid T cells. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay | |
KP1 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy | |
CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1 | |
Mouse | |
Protein A or G purified | |
Cell Biology, Cytokine Research, Immunology, Microglia Markers | |
968 | |
Human, Mouse, Rat | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction