Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD68/SR-D1 Mouse, Clone: C68/684, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $536.00
Specifications
Antigen | CD68/SR-D1 |
---|---|
Clone | C68/684 |
Dilution | Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP232832F
![]() |
Novus Biologicals
NBP2328320.02MG |
0.02 mg |
Each for $268.00
|
|
|||||
NBP232832A
![]() |
Novus Biologicals
NBP2328320.1MG |
0.1 mg |
Each for $536.00
|
|
|||||
NBP232832B
![]() |
Novus Biologicals
NBP2328320.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
CD68/SR-D1 | |
Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Feline, Primate, Rat (Negative) | |
P34810 | |
968 | |
Subcellular fraction of human alveolar macrophages | |
Primary | |
Store at 4C. | |
110 kDa |
C68/684 | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cell Biology, Cytokine Research, Immunology | |
PBS with 0.05% BSA. with 0.05% Sodium Azide | |
CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1 | |
CD68 | |
IgG1 κ | |
Protein A purified | |
This antibody recognizes a glycoprotein of 110kDa, which is identified as CD68. It is important for identifying macrophages in tissue sections. It stains macrophages in a wide variety of human tissues, including Kupffer cells and macrophages in the red pulp of the spleen, in lamina propria of the gut, in lung alveoli, and in bone marrow. It reacts with myeloid precursors and peripheral blood granulocytes. It also reacts with plasmacytoid T cells, which are supposed to be of monocyte/macrophage origin. It shows strong granular cytoplasmic staining of chronic and acute myeloid leukemia and also reacts with rare cases of true histiocytic neoplasia. Lymphomas are negative or show few granules. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title