Learn More
CD69 Antibody (15B5G2), Alexa Fluor™ 594, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP225236AF594
Catalog No. NP22236A94
Only null left
Add to Cart
Description
Description
CD69 Monoclonal specifically detects CD69 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
CD69
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
CD69
Amino acids 103-199 of human CD69
Primary
Human
IgG2a κ
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
15B5G2
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leukocyte surface antigen Leu-23, MLR-3
Mouse
0.1 mL
969
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.