Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD69 Antibody (30H2), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | CD69 |
---|---|
Clone | 30H2 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
CD69 Monoclonal antibody specifically detects CD69 in Human samples. It is validated for ELISASpecifications
CD69 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leukocyte surface antigen Leu-23, MLR-3 | |
Recombinant Human CD69 protein [UniProt Q07108] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
30H2 | |
ELISA | |
Unconjugated | |
Human | |
Innate Immunity | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
969 | |
IgG4 | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title