Learn More
Description
Specifications
Specifications
| Antigen | CD69 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | FN50 |
| Conjugate | FITC |
| Dilution | Flow Cytometry 1:10-1:1000 |
| Formulation | Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide |
| Gene Alias | Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leukocyte surface antigen Leu-23, MLR-3 |
| Gene Symbols | CD69 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
