Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD69 Antibody (H1.2F3), Alexa Fluor™ 405, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP128011AF405
Description
CD69 Monoclonal specifically detects CD69 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD69 | |
Monoclonal | |
Alexa Fluor 405 | |
Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leukocyte surface antigen Leu-23, MLR-3 | |
Armenian Hamster | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse CD69/Very Early Activation Antigen (VEA), Mr 85 kDa (unreduced) | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
H1.2F3 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
CD69 | |
Mouse dendritic epidermal cell line Y245 | |
0.25 mL | |
Innate Immunity | |
969 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction