Learn More
CD69 Antibody (H1.2F3), DyLight 350, Novus Biologicals™
SDP
Armenian Hamster Monoclonal Antibody
Supplier:  Novus Biologicals NBP128011UV
Catalog No. NBP128011UV
Only null left
Add to Cart
Description
Description
CD69 Monoclonal specifically detects CD69 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
CD69
Monoclonal
DyLight 350
Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leukocyte surface antigen Leu-23, MLR-3
Armenian Hamster
Protein A or G purified
RUO
Primary
Mouse CD69/Very Early Activation Antigen (VEA), Mr 85 kDa (unreduced)
Store at 4C in the dark.
IgG1
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
H1.2F3
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
CD69
Mouse dendritic epidermal cell line Y245
0.25 mL
Innate Immunity
969
Mouse
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only