Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD69 Armenian Hamster anti-Mouse, FITC, Clone: H1.2F3, Proteintech
SDP

Catalog No. 502258253 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-225-8253 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-225-8253 Supplier Proteintech Group Inc Supplier No. FITC65105100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Armenian Hamster Monoclonal Antibody

The CD69 antibody from Proteintech is a armenian hamster monoclonal antibody generated with mouse dendritic epidermal t cell line y245 of mouse CD69. This antibody recognizes mouse antigen. The CD69 antibody has been validated for the following applications: FC analysis. This antibody is fitc.

Specifications

Antigen CD69
Applications Flow Cytometry
Classification Monoclonal
Clone H1.2F3
Concentration 0.5 mg/mL
Conjugate FITC
Host Species Armenian Hamster
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 12515
Target Species Mouse
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.