Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd69, Hamster anti-Mouse, FITC, Clone: H1.2F3, Abnova™

Catalog No. 89998924 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-924 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-924 Supplier Abnova Corporation Supplier No. MAB16150
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

Specifications

Antigen Cd69
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone H1.2F3
Conjugate FITC
Dilution Flow Cytometry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunoprecipitation The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide).
Gene CD69 antigen
Gene Alias 5830438K24Rik/AI452015/AIM/VEA
Gene Symbols Cd69
Host Species Hamster
Immunogen Y245 mouse dendritic epidermal T cell line.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12515
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C. Avoid prolonged exposure to light and repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.