Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD69 Hamster anti-Mouse, PE, Clone: H1.2F3, BD

Catalog No. BDB555318 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
25 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555318 0.5 mg
BDB561932 25 μg
BDB561941 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BDB555318 Supplier BD Biosciences Supplier No. 555318
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The In-1 monoclonal antibody specifically binds to CD74, the MHC class II-associated Invariant chain (Ii), expressed in I-A+ cell populations. Although it is predominantly expressed intracellularly, it has also been detected on the cell surface of some B-cell lines and at low levels on a small number of B lymphocytes. In the mouse, alternatively spliced transcripts encode 31-kDa and 41-kDa isoforms, and the cell-surface CD74 has a chondroitin sulfate side chain which allows it to interact with CD44. CD74 is directly involved in the intracellular transport of MHC class II molecules and antigen presentation, and it affects the maturation of T and B lymphocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD74
Applications Flow Cytometry
Classification Monoclonal
Clone IN-1
Concentration 0.5mg/mL
Conjugate FITC
Description Cd74; CLIP; Ii; Ia-associated invariant chain; Ia-GAMMA; DHLAG; HG2A; HLADG
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen ATL mouse lymphocytes and B10A mouse-derived CH1 B-cell lymphoma
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.