Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD69 Mouse anti-Baboon,Cynomolgus,Human,Rhesus, Brilliant Ultraviolet 737, Clone: FN50, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 612818
Description
Applications:
Flow cytometry

Specifications
CD69 | |
Monoclonal | |
Brilliant Ultraviolet 737 | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
FN50 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
IgG1 κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction