Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD69 Mouse anti-Human, BUV805, Clone: FN50 (also known as FN 50), BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569163
Description
CD69 Monoclonal antibody specifically detects CD69 in Human samples. It is validated for Flow cytometry
Specifications
CD69 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
969 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG1 κ |
Flow Cytometry | |
FN50 (also known as FN 50) | |
Brilliant Ultraviolet 805 | |
AIM; CLEC2C; EA1; GP32/28; Leu23; MLR-3; VEA; BL-AC/P26 | |
Anti-μ stimulated human B lymphocytes | |
50 μg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction