Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD69 Mouse anti-Human, Unconjugated, Clone: FN61, MilliporeSigma™

Catalog No. MABF22102MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF22102MI 25 μg
MABF22101MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF22102MI Supplier MilliporeSigma Supplier No. MABF221025UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD69 Monoclonal antibody specifically detects CD69 in Human samples. It is validated for Combined Interference Assay, Flow Cytometry, Inhibits Activity/Function, Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD69
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone FN61
Conjugate Unconjugated
Gene Accession No. NP_001772.1
Gene Alias Early activation antigen CD69;Activation inducer molecule;AIM;BL-AC/P26;C-type lectin domain family 2 member C;EA1;Early T-cell activation antigen p60;GP32/28;Leukocyte surface antigen Leu-23;MLR-3
Gene Symbols CD69, CLEC2C
Host Species Mouse
Immunogen Activated B cells from human peripheral blood.
Molecular Weight of Antigen 22.56 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone FN61 is a mouse monoclonal antibody that detects human Early activation antigen CD69. It targets an epitope within the E1 site in the extracellular domain.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.