Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD7 Antibody (017), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290559AF647
Description
CD7 Monoclonal specifically detects CD7 in Mouse samples. It is validated for ELISA.Specifications
CD7 | |
Monoclonal | |
Alexa Fluor 647 | |
CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD7 extracellular domain (NP_033984.1; Met 1-Pro 150). | |
0.1 mL | |
Cytokine Research, Signal Transduction | |
924 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
17 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction