Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD7 Antibody (15), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289748MFV450
Description
CD7 Monoclonal antibody specifically detects CD7 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISASpecifications
CD7 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse (Negative), Rat (Negative) | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
15 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA Capture | |
CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD7 (Accession#: NP_006128.1; Met 1-Pro 180). | |
0.1 mL | |
Cytokine Research, Signal Transduction | |
924 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction