Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD7 Antibody (3A1E), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281050H
Description
CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Functional AssaySpecifications
| CD7 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Functional Assay | |
| 3A1E | |
| Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Functional | |
| CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein | |
| CD7 | |
| 0.1 mL | |
| Cytokine Research, Signal Transduction | |
| 924 | |
| Store at 4C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction