Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD7 Antibody (3A1E), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281050JF646
Description
CD7 Monoclonal specifically detects CD7 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
CD7 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
CD7 | |
Recombinant monoclonal antibody to CD7. | |
0.1 mL | |
Cytokine Research, Signal Transduction | |
924 | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
3A1E | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Recognises human CD7. | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction