Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD7 Antibody (V55P2F2*B12), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250142AF532
Description
CD7 Monoclonal specifically detects CD7 in Fish samples. It is validated for ELISA.Specifications
CD7 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
CD7 | |
Ovalbumin-conjugated synthetic peptide | |
0.1 mL | |
924 | |
Store at 4°C in the dark. |
ELISA | |
V55P2F2*B12 | |
ELISA | |
CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein | |
Mouse | |
Protein A purified | |
Primary | |
Fish | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction