Learn More
CD7 Monoclonal Antibody (MG34), FITC, Exalpha

Mouse Monoclonal Antibody
Supplier: Vector Laboratories 0072100TESTS


Description
Flow Cytometry PBMC: Add 10 μL of MAB/10^6 PBMC in 100 μL PBS. Mix gently and incubate for 15 minutes at 2 to 8C. Wash twice with PBS and analyze or fix with 0.5% v/v of paraformaldehyde in PBS and analyze. Flow Cytometry Whole Blood: Add10 μL of MAB/100 μL of whole blood. Mix gently and incubate for 15 minutes at room temperature 20C. Lyse the whole blood. Wash once with PBS and analyze or fix with 0.5% v/v of paraformaldehyde in PBS and analyze. See instrument manufacturer instructions for Lysed Whole Blood and Immunofluorescence analysis with a flow cytometer or microscope.
CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Flow CytometrySpecifications
CD7 | |
Monoclonal | |
FITC | |
CD7 | |
Cd7; CD7 antigen; CD7 antigen (p41); Cd7 molecule; GP40; LEU-9; p41 protein; T-cell antigen CD7; T-cell leukemia antigen; T-cell surface antigen Leu-9; Tp40; TP41 | |
CD7 was derived from the hybridization of mouse P3-X63-Ag8.653 myeloma cells with spleen cells of BALB/c mice immunized with T-acute lymphoblastic leukemia (T-ALL) cells. | |
100Tests | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry | |
MG34 | |
PBS with 0.075% sodium azide; pH 7.2 | |
P09564 | |
Mouse | |
Protein A/G | |
RUO | |
924 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.