Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD7 Mouse anti-Human, BV421, Clone: 4H9 (also known as Leu-9; Leu9), BD Horizon™
Mouse Monoclonal Antibody
$178.00 - $434.00
Specifications
Antigen | CD7 |
---|---|
Clone | 4H9 (also known as Leu-9; Leu9) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Violet 421 |
Description
CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Flow cytometry (Routinely Tested)Specifications
CD7 | |
Flow Cytometry | |
Brilliant Violet 421 | |
Mouse | |
Human | |
LEU-9; T-cell leukemia antigen; T-cell surface antigen Leu-9 | |
T-acute lymphoblastic leukemia cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
4H9 (also known as Leu-9; Leu9) | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
924 | |
IgG2a κ | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title