Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD70 Monoclonal Antibody (TAN1-7), Functional Grade, In Vivo GOLD™, Leinco Technologies™
SDP

Catalog No. 502544149
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4149 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4149 Supplier Leinco Technologies Inc Supplier No. I2002100MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specificity: Clone TAN1-7 monoclonal antibody recognizes a direct epitope on mouse CD70 and capable of blocking the CD70-CD27 interaction involved in B and T cell activation. Endotoxin Level: <1.0 EU/mg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.

CD70 Monoclonal antibody specifically detects CD70 in Mouse samples. It is validated for ELISA, Flow Cytometry, Functional Assay, Immunocytochemistry, Inhibition Assays

Specifications

Antigen CD70
Applications ELISA, Flow Cytometry, Functional Assay, Immunocytochemistry, Inhibition Assays
Classification Monoclonal
Clone TAN1-7
Concentration ≥5.0 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2-7.4
Gene Cd70
Gene Accession No. O55237
Gene Alias CD27 ligand; Cd27l; CD27-L; CD27LG; CD70; CD70 antigen; Cd70 molecule; Ki-24 antigen; surface antigen CD70; Tnfsf7; TNLG8A; tumor necrosis factor (ligand) superfamily, member 7; tumor necrosis factor ligand 8A; tumor necrosis factor ligand superfamily member 7
Host Species Mouse
Immunogen Recombinant mouse CD70 (extracellular domain, aa residues 41-195)
Purification Method Protein A/G
Quantity 100 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21948
Target Species Mouse
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.